Product Details
- SNP ID
-
hCV30439897
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:83878257 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGACCCAGAGTTGAAAACATTTTCG[C/T]TTTAATACTGAAAAAATATGCCATT
- Phenotype
-
MIM: 609678
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLITRK1
PubMed Links
Gene Details
- Gene
- SLITRK1
- Gene Name
- SLIT and NTRK like family member 1
There are no transcripts associated with this gene.
View Full Product Details