Product Details

SNP ID
rs7556338
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:15531723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGGAATAATTCATCACCATAGAA[A/G]TGTATTGAGTGTGTCATCCAGTAAA
Phenotype
MIM: 602234
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASP9 PubMed Links

Gene Details

Gene
CASP9
Gene Name
caspase 9
There are no transcripts associated with this gene.

Gene
DNAJC16
Gene Name
DnaJ heat shock protein family (Hsp40) member C16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287811.1 Intron NP_001274740.1
NM_015291.3 Intron NP_056106.1

View Full Product Details