Product Details

SNP ID
rs9967846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:132644651 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTCCATTTGATTTAAAAATATCA[C/G]TAGGTCTTCTTTGTGCAGACATCCT
Phenotype
MIM: 602886 MIM: 610450
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPR39 PubMed Links

Gene Details

Gene
GPR39
Gene Name
G protein-coupled receptor 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001508.2 2169 Intron NP_001499.1
XM_011511021.2 2169 Intron XP_011509323.1
Gene
LYPD1
Gene Name
LY6/PLAUR domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077427.3 2169 UTR 3 NP_001070895.1
NM_001321234.1 2169 UTR 3 NP_001308163.1
NM_001321235.1 2169 UTR 3 NP_001308164.1
NM_144586.6 2169 UTR 3 NP_653187.3
XM_011510546.2 2169 UTR 3 XP_011508848.1

View Full Product Details