Product Details

SNP ID
rs10499045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:106637699 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACCTAGTTTCCATTAGAGAAAAGA[C/T]GTGCCAGAGAGAGAATTTTTTTTCT
Phenotype
MIM: 610502
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
QRSL1 PubMed Links

Gene Details

Gene
QRSL1
Gene Name
glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018292.4 Intron NP_060762.3
XM_011535924.2 Intron XP_011534226.1
Gene
RTN4IP1
Gene Name
reticulon 4 interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details