Product Details

SNP ID
rs6535556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:147744055 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTGTGAGAAGAAACGTGTATATT[A/G]GTGCAGAAGAGTTCTAGTGCAGGAG
Phenotype
MIM: 609746
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP10 PubMed Links

Gene Details

Gene
ARHGAP10
Gene Name
Rho GTPase activating protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024605.3 Intron NP_078881.3
XM_005263215.3 Intron XP_005263272.1
XM_017008602.1 Intron XP_016864091.1

View Full Product Details