Product Details

SNP ID
rs9832065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:33296527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGCTCTTATATTTATTTAAGTC[A/G]CTGAGCCATTTTGAATTTTTTTTTA
Phenotype
MIM: 605652
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXL2 PubMed Links

Gene Details

Gene
FBXL2
Gene Name
F-box and leucine rich repeat protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171713.1 Intron NP_001165184.1
NM_012157.3 Intron NP_036289.3
XM_005265014.2 Intron XP_005265071.1
XM_005265015.3 Intron XP_005265072.1
XM_005265016.2 Intron XP_005265073.1
XM_005265017.3 Intron XP_005265074.1
XM_011533559.2 Intron XP_011531861.1
XM_017006092.1 Intron XP_016861581.1
XM_017006093.1 Intron XP_016861582.1
XM_017006094.1 Intron XP_016861583.1

View Full Product Details