Product Details

SNP ID
rs6701389
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:182413689 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAACTGGCCAGCGTTTGCTTTTG[A/T]TGACAGCGAACCCTCAGATCTAGTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00272 PubMed Links

Gene Details

Gene
LINC00272
Gene Name
long intergenic non-protein coding RNA 272
There are no transcripts associated with this gene.

View Full Product Details