Product Details

SNP ID
rs3093457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:155997942 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCTCCTCTGGTGTCCTGGCCTTG[G/T]GTATCTCTTAGAGATGGCAAAATTT
Phenotype
MIM: 300007
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IL9R PubMed Links

Gene Details

Gene
IL9R
Gene Name
interleukin 9 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002186.2 Intron NP_002177.2
NM_176786.1 Intron NP_789743.2
XM_011531151.2 Intron XP_011529453.1
XM_011531152.2 Intron XP_011529454.1
XM_011531154.2 Intron XP_011529456.1
XM_011531155.2 Intron XP_011529457.1
XM_011531156.2 Intron XP_011529458.1
XM_011531157.2 Intron XP_011529459.1
XM_017029495.1 Intron XP_016884984.1
XM_017029496.1 Intron XP_016884985.1
XM_017029497.1 Intron XP_016884986.1
XM_017029498.1 Intron XP_016884987.1
XM_017029499.1 Intron XP_016884988.1
XM_017029500.1 Intron XP_016884989.1
XM_017029501.1 Intron XP_016884990.1
XM_017029502.1 Intron XP_016884991.1
XM_017029503.1 Intron XP_016884992.1
XM_017029504.1 Intron XP_016884993.1
XM_017029505.1 Intron XP_016884994.1
XM_017029506.1 Intron XP_016884995.1

View Full Product Details