Product Details

SNP ID
rs6062426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63258407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGTGATGCCATTTAGGGCCCAC[C/T]TATGTAATCCAGCATGCTCTCATGT
Phenotype
MIM: 612873
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLJ16779 PubMed Links

Gene Details

Gene
FLJ16779
Gene Name
uncharacterized LOC100192386
There are no transcripts associated with this gene.

Gene
NKAIN4
Gene Name
Na+/K+ transporting ATPase interacting 4
There are no transcripts associated with this gene.

View Full Product Details