Product Details

SNP ID
rs9641226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:98108067 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAAAAATCTAGGCTAACTTGGC[A/G]TTATTGACTGCCGCCATTACCTTTG
Phenotype
MIM: 610989
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LMTK2 PubMed Links

Gene Details

Gene
LMTK2
Gene Name
lemur tyrosine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014916.3 Intron NP_055731.2
XM_011515981.2 Intron XP_011514283.1

View Full Product Details