Product Details

SNP ID
rs10484591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:25656433 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCAACCAGCCTGCTTGTAATTAC[C/G]TATTCCTCAGAGCCTCGGAGGGGAT
Phenotype
MIM: 609202
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SCGN PubMed Links

Gene Details

Gene
SCGN
Gene Name
secretagogin, EF-hand calcium binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006998.3 Intron NP_008929.2
XM_017010181.1 Intron XP_016865670.1

View Full Product Details