Product Details

SNP ID
rs5970449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153342189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATGTCATCAAGTTCTCTAATTTT[C/T]TTGTGTGGTATCTAATCTGCTGTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105373378 PubMed Links

Gene Details

Gene
LOC105373378
Gene Name
uncharacterized LOC105373378
There are no transcripts associated with this gene.

Gene
ZNF275
Gene Name
zinc finger protein 275
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080485.3 Intron NP_001073954.3

View Full Product Details