Product Details

SNP ID
rs10408073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9756837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATGAAGTACTATGAAGGTTTGTG[A/G]AGTATTTTAAGGCTATCTCTCATTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF846 PubMed Links

Gene Details

Gene
ZNF846
Gene Name
zinc finger protein 846
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077624.1 2729 Intron NP_001071092.1
XM_005259772.3 2729 UTR 3 XP_005259829.1
XM_005259773.4 2729 UTR 3 XP_005259830.1
XM_005259775.3 2729 UTR 3 XP_005259832.1
XM_006722658.3 2729 Intron XP_006722721.1
XM_011527717.2 2729 UTR 3 XP_011526019.1
XM_011527718.2 2729 UTR 3 XP_011526020.1
XM_011527719.2 2729 UTR 3 XP_011526021.1
XM_011527720.2 2729 UTR 3 XP_011526022.1
XM_011527721.2 2729 UTR 3 XP_011526023.1
XM_017026404.1 2729 UTR 3 XP_016881893.1
XM_017026405.1 2729 Intron XP_016881894.1
XM_017026406.1 2729 Intron XP_016881895.1
XM_017026407.1 2729 Intron XP_016881896.1
XM_017026408.1 2729 Intron XP_016881897.1

View Full Product Details