Product Details

SNP ID
rs6690028
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:247915442 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTCCATGTGACTGCCCCCCTGCT[C/T]TGTGCTACAAGAACACATCTGTGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2T8 PubMed Links

Gene Details

Gene
OR2T8
Gene Name
olfactory receptor family 2 subfamily T member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005522.1 1305 Intron NP_001005522.1
XM_011544178.2 1305 Intron XP_011542480.1
XM_017001172.1 1305 UTR 5 XP_016856661.1

View Full Product Details