Product Details

SNP ID
rs6442122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48456895 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAAATGAGTAAAAGAATTTTCAA[C/T]AATTTGGATCAAAATCCCTGGAATT
Phenotype
MIM: 606605 MIM: 606609
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATRIP PubMed Links

Gene Details

Gene
ATRIP
Gene Name
ATR interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271022.1 Intron NP_001257951.1
NM_001271023.1 Intron NP_001257952.1
NM_032166.3 Intron NP_115542.2
NM_130384.2 Intron NP_569055.1
Gene
TREX1
Gene Name
three prime repair exonuclease 1
There are no transcripts associated with this gene.

View Full Product Details