Product Details

SNP ID
rs9991222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:119223208 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCAATATTTTGAGTTGTTCCTTT[A/G]TTTGGCACCTGGAGATTTTTGTAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
USP53 PubMed Links

Gene Details

Gene
USP53
Gene Name
ubiquitin specific peptidase 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019050.2 Intron NP_061923.2
XM_005263073.3 Intron XP_005263130.1
XM_005263075.2 Intron XP_005263132.1
XM_005263076.2 Intron XP_005263133.1
XM_005263077.3 Intron XP_005263134.1
XM_006714240.1 Intron XP_006714303.1
XM_011532037.2 Intron XP_011530339.1
XM_011532038.2 Intron XP_011530340.1
XM_017008312.1 Intron XP_016863801.1
XM_017008313.1 Intron XP_016863802.1
XM_017008314.1 Intron XP_016863803.1
XM_017008315.1 Intron XP_016863804.1
XM_017008316.1 Intron XP_016863805.1
XM_017008317.1 Intron XP_016863806.1
XM_017008318.1 Intron XP_016863807.1

View Full Product Details