Product Details
- SNP ID
-
rs10100467
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:29333707 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACACCCGGGGAGCTACGAAACACAC[G/T]GATGCCTGAGTCCCCCACCTAGAAT
- Phenotype
-
MIM: 602747
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DUSP4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112637547] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DUSP4
- Gene Name
- dual specificity phosphatase 4
View Full Product Details