Product Details

SNP ID
rs7535470
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:32252121 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAAGTTGGAAGTGAAAGAGAATG[A/G]TGGGAGAAAGGAAGAGGAAGTACTA
Phenotype
MIM: 153390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM167B PubMed Links

Gene Details

Gene
FAM167B
Gene Name
family with sequence similarity 167 member B
There are no transcripts associated with this gene.

Gene
LCK
Gene Name
LCK proto-oncogene, Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042771.2 Intron NP_001036236.1
NM_005356.4 Intron NP_005347.3
XM_011541453.2 Intron XP_011539755.1
XM_011541454.2 Intron XP_011539756.1
XM_011541455.2 Intron XP_011539757.1

View Full Product Details