Product Details
- SNP ID
-
rs13432480
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:231924305 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGGTTCACAAACTGTGAAAAAGTG[C/T]TCTGTACTGTAAAAAAGAGACAGTG
- Phenotype
-
MIM: 600296
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NPPC
PubMed Links
Gene Details
- Gene
- NPPC
- Gene Name
- natriuretic peptide C
View Full Product Details