Product Details

SNP ID
rs12471298
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96861051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGCCTTGGCCCCTTTCTCCAGCT[A/C]TTCCCGCAGCCGCCGGCGCAATGAC
Phenotype
MIM: 604462
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
ANKRD39 PubMed Links
Additional Information
For this assay, SNP(s) [rs12476284] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ANKRD39
Gene Name
ankyrin repeat domain 39
There are no transcripts associated with this gene.

Gene
SEMA4C
Gene Name
semaphorin 4C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017789.4 2237 Nonsense Mutation GAG,TAG E693* NP_060259.4
XM_006712606.3 2237 Intron XP_006712669.2
XM_011511378.2 2237 Nonsense Mutation GAG,TAG E693* XP_011509680.1
XM_011511379.2 2237 Nonsense Mutation GAG,TAG E693* XP_011509681.1
XM_011511380.1 2237 Nonsense Mutation GAG,TAG E693* XP_011509682.1
XM_011511381.1 2237 Nonsense Mutation GAG,TAG E693* XP_011509683.1
XM_011511382.2 2237 Nonsense Mutation GAG,TAG E693* XP_011509684.1
XM_011511383.1 2237 Nonsense Mutation GAG,TAG E693* XP_011509685.1
XM_017004393.1 2237 Nonsense Mutation GAG,TAG E735* XP_016859882.1
XM_017004394.1 2237 Nonsense Mutation GAG,TAG E552* XP_016859883.1

View Full Product Details