Product Details

SNP ID
rs10875952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49380777 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTTCCACCTTTGCCTACTGTGA[A/G]GAAGGTTACTGTGAACATTCATGTA
Phenotype
MIM: 611667
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100335030 PubMed Links

Gene Details

Gene
LOC100335030
Gene Name
FGFR1 oncogene partner 2 pseudogene
There are no transcripts associated with this gene.

Gene
SPATS2
Gene Name
spermatogenesis associated serine rich 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293285.1 Intron NP_001280214.1
NM_001293286.1 Intron NP_001280215.1
NM_023071.3 Intron NP_075559.2
XM_011538690.2 Intron XP_011536992.1
XM_011538691.2 Intron XP_011536993.1
XM_017019840.1 Intron XP_016875329.1

View Full Product Details