Product Details
- SNP ID
-
rs11780870
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:91074864 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTACTGTTATACTTTGGGTACGG[G/T]TCTTCTTTAAGCCTCAGTTTCACAT
- Phenotype
-
MIM: 612021
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OTUD6B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs140038684,rs144533375] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OTUD6B
- Gene Name
- OTU domain containing 6B
- Gene
- OTUD6B-AS1
- Gene Name
- OTUD6B antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details