Product Details

SNP ID
rs11107940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:95221227 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTATTTACATACACTGGATACAAG[G/T]CCTTTGTTAGAGATGCAGTTGGCCC
Phenotype
MIM: 613520
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FGD6 PubMed Links

Gene Details

Gene
FGD6
Gene Name
FYVE, RhoGEF and PH domain containing 6
There are no transcripts associated with this gene.

Gene
VEZT
Gene Name
vezatin, adherens junctions transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017599.3 Intron NP_060069.3
XM_006719477.3 Intron XP_006719540.1
XM_006719478.2 Intron XP_006719541.1
XM_006719479.2 Intron XP_006719542.1
XM_006719480.3 Intron XP_006719543.1
XM_006719481.2 Intron XP_006719544.1
XM_006719482.3 Intron XP_006719545.1
XM_006719483.3 Intron XP_006719546.1
XM_006719484.3 Intron XP_006719547.1
XM_006719485.2 Intron XP_006719548.1
XM_006719486.3 Intron XP_006719549.1
XM_006719490.3 Intron XP_006719553.1
XM_011538543.1 Intron XP_011536845.1
XM_011538544.1 Intron XP_011536846.1
XM_011538545.2 Intron XP_011536847.1
XM_011538546.1 Intron XP_011536848.1
XM_011538547.1 Intron XP_011536849.1
XM_011538548.2 Intron XP_011536850.1
XM_011538549.1 Intron XP_011536851.1
XM_011538550.2 Intron XP_011536852.1
XM_011538551.2 Intron XP_011536853.1
XM_011538552.1 Intron XP_011536854.1
XM_011538553.2 Intron XP_011536855.1
XM_017019586.1 Intron XP_016875075.1
XM_017019587.1 Intron XP_016875076.1
XM_017019588.1 Intron XP_016875077.1
XM_017019589.1 Intron XP_016875078.1
XM_017019590.1 Intron XP_016875079.1
XM_017019591.1 Intron XP_016875080.1
XM_017019592.1 Intron XP_016875081.1
XM_017019593.1 Intron XP_016875082.1
XM_017019594.1 Intron XP_016875083.1
XM_017019595.1 Intron XP_016875084.1
XM_017019596.1 Intron XP_016875085.1
XM_017019597.1 Intron XP_016875086.1
XM_017019598.1 Intron XP_016875087.1
XM_017019599.1 Intron XP_016875088.1
XM_017019600.1 Intron XP_016875089.1
XM_017019601.1 Intron XP_016875090.1
XM_017019602.1 Intron XP_016875091.1
XM_017019603.1 Intron XP_016875092.1
XM_017019604.1 Intron XP_016875093.1
XM_017019605.1 Intron XP_016875094.1

View Full Product Details