Product Details
- SNP ID
-
rs4889651
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:30884224 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCCTAGGCCCCGTGACCAGGAGAG[G/A]GTCAAGAATGGGGGAGACTCAAACT
- Phenotype
-
MIM: 605847
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BCL7C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78340624] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BCL7C
- Gene Name
- BCL tumor suppressor 7C
- Gene
- MIR4519
- Gene Name
- microRNA 4519
There are no transcripts associated with this gene.
- Gene
- MIR762
- Gene Name
- microRNA 762
There are no transcripts associated with this gene.
- Gene
- MIR762HG
- Gene Name
- MIR762 host gene
There are no transcripts associated with this gene.
View Full Product Details