Product Details

SNP ID
rs11572951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123644913 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTGTTATTCTTTAAATGTTACAC[G/T]GCAAATATTTGTGGAATATTGTACT
Phenotype
MIM: 601750
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GTF2H3 PubMed Links

Gene Details

Gene
GTF2H3
Gene Name
general transcription factor IIH subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271866.1 Intron NP_001258795.1
NM_001271867.1 Intron NP_001258796.1
NM_001271868.1 Intron NP_001258797.1
NM_001516.4 Intron NP_001507.2
XM_017019228.1 Intron XP_016874717.1

View Full Product Details