Product Details

SNP ID
rs13282494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:53852660 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAAAAAATACAGTTTGAGTCTG[G/T]AAATCCTCTTAGCTATTAATAGAAC
Phenotype
MIM: 608861 MIM: 607193
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATP6V1H PubMed Links

Gene Details

Gene
ATP6V1H
Gene Name
ATPase H+ transporting V1 subunit H
There are no transcripts associated with this gene.

Gene
RGS20
Gene Name
regulator of G-protein signaling 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286673.1 Intron NP_001273602.1
NM_001286674.1 Intron NP_001273603.1
NM_001286675.1 Intron NP_001273604.1
NM_003702.4 Intron NP_003693.2
NM_170587.3 Intron NP_733466.1
XM_011517622.2 Intron XP_011515924.1

View Full Product Details