Product Details

SNP ID
rs12972871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14032319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCAATTGTCAGAAGTCCAAGTT[C/T]TGGGGTGTGCAGGCGGAAGGGGGGG
Phenotype
MIM: 605350 MIM: 606855
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL27RA PubMed Links

Gene Details

Gene
IL27RA
Gene Name
interleukin 27 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004843.3 Intron NP_004834.1
Gene
RLN3
Gene Name
relaxin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001311197.1 Intron NP_001298126.1
NM_080864.3 Intron NP_543140.1

View Full Product Details