Product Details

SNP ID
rs13032663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216085530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTTAAGAGACAGTGGCCTCCTA[G/T]ACCAGAGGGGAAGCAGTGGAGATGG
Phenotype
MIM: 605843
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PECR PubMed Links

Gene Details

Gene
PECR
Gene Name
peroxisomal trans-2-enoyl-CoA reductase
There are no transcripts associated with this gene.

Gene
TMEM169
Gene Name
transmembrane protein 169
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142310.1 Intron NP_001135782.1
NM_001142311.1 Intron NP_001135783.1
NM_001142312.1 Intron NP_001135784.1
NM_138390.3 Intron NP_612399.1
XM_006712851.1 Intron XP_006712914.1
XM_011512173.2 Intron XP_011510475.1

View Full Product Details