Product Details

SNP ID
rs13030384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:25929052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCATGTGTGGTGGAAGGTGGAG[C/G]CCGCTGAGGACTCTGGCACCTGCAG
Phenotype
MIM: 602845
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIF3C PubMed Links

Gene Details

Gene
KIF3C
Gene Name
kinesin family member 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002254.6 2962 Missense Mutation CCT,GCT P770A NP_002245.4
XM_005264299.2 2962 Missense Mutation CCT,GCT P769A XP_005264356.2

View Full Product Details