Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001080494.3 | 2566 | UTR 3 | NP_001073963.1 | ||
NM_001144832.2 | 2566 | UTR 3 | NP_001138304.1 | ||
NM_001297662.1 | 2566 | Intron | NP_001284591.1 | ||
NM_001297663.1 | 2566 | UTR 3 | NP_001284592.1 | ||
NM_001297664.1 | 2566 | UTR 3 | NP_001284593.1 | ||
NM_001297665.1 | 2566 | UTR 3 | NP_001284594.1 | ||
NM_001297666.1 | 2566 | UTR 3 | NP_001284595.1 | ||
NM_001297667.1 | 2566 | UTR 3 | NP_001284596.1 | ||
XM_005270644.2 | 2566 | UTR 3 | XP_005270701.1 | ||
XM_006710471.2 | 2566 | UTR 3 | XP_006710534.1 | ||
XM_011541048.2 | 2566 | UTR 3 | XP_011539350.1 | ||
XM_011541049.2 | 2566 | UTR 3 | XP_011539351.1 | ||
XM_011541050.2 | 2566 | UTR 3 | XP_011539352.1 | ||
XM_011541051.2 | 2566 | UTR 3 | XP_011539353.1 | ||
XM_011541052.2 | 2566 | UTR 3 | XP_011539354.1 | ||
XM_011541053.2 | 2566 | UTR 3 | XP_011539355.1 | ||
XM_011541054.2 | 2566 | UTR 3 | XP_011539356.1 | ||
XM_017000715.1 | 2566 | UTR 3 | XP_016856204.1 |