Product Details

SNP ID
rs11205997
Assay Type
Functionally tested
NCBI dbSNP Submissions
42
Location
Chr.1:52729599 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATTTGTTAAATGTTAATTAGGTT[C/G]TGTTTGAATAGTTGAGATGTTTAAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZYG11B PubMed Links

Gene Details

Gene
ZYG11B
Gene Name
zyg-11 family member B, cell cycle regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024646.2 Intron NP_078922.1
XM_006710898.3 Intron XP_006710961.1
XM_017002336.1 Intron XP_016857825.1

View Full Product Details