Product Details
- SNP ID
-
rs12026551
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:182850854 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTTTCAATACTCTTATACCTCAC[C/G]TATTTTAATTGATTTTGAAATGTTA
- Phenotype
-
MIM: 603115
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DHX9
PubMed Links
Gene Details
- Gene
- DHX9
- Gene Name
- DEAH-box helicase 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001357.4 |
|
Intron |
|
|
NP_001348.2 |
View Full Product Details