Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_006377.3 | Intron | NP_006368.3 | |||
XM_011517681.2 | Intron | XP_011515983.1 | |||
XM_011517682.2 | Intron | XP_011515984.1 | |||
XM_011517683.2 | Intron | XP_011515985.1 | |||
XM_011517684.2 | Intron | XP_011515986.1 | |||
XM_011517685.2 | Intron | XP_011515987.1 | |||
XM_011517686.2 | Intron | XP_011515988.1 | |||
XM_017014191.1 | Intron | XP_016869680.1 | |||
XM_017014192.1 | Intron | XP_016869681.1 |