Product Details
- SNP ID
-
rs11855870
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:72199707 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAAGCCGGAGCCAGGGCGCCATCC[A/G]GTCAGCACAATGACCACATCTCCCT
- Phenotype
-
MIM: 179050
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GRAMD2
PubMed Links
Gene Details
- Gene
- GRAMD2
- Gene Name
- GRAM domain containing 2
There are no transcripts associated with this gene.
- Gene
- PKM
- Gene Name
- pyruvate kinase, muscle
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001206796.2 |
2061 |
Silent Mutation |
ACC,ACT |
T587T |
NP_001193725.1 |
NM_001206797.2 |
2061 |
Silent Mutation |
ACC,ACT |
T439T |
NP_001193726.1 |
NM_001206798.2 |
2061 |
Silent Mutation |
ACC,ACT |
T498T |
NP_001193727.1 |
NM_001206799.1 |
2061 |
Intron |
|
|
NP_001193728.1 |
NM_001316318.1 |
2061 |
Silent Mutation |
ACC,ACT |
T548T |
NP_001303247.1 |
NM_002654.5 |
2061 |
Silent Mutation |
ACC,ACT |
T513T |
NP_002645.3 |
NM_182470.3 |
2061 |
Silent Mutation |
ACC,ACT |
T513T |
NP_872270.1 |
NM_182471.3 |
2061 |
Silent Mutation |
ACC,ACT |
T513T |
NP_872271.1 |
XM_005254443.1 |
2061 |
Silent Mutation |
ACC,ACT |
T573T |
XP_005254500.1 |
XM_005254445.4 |
2061 |
Intron |
|
|
XP_005254502.1 |
XM_006720570.1 |
2061 |
Silent Mutation |
ACC,ACT |
T587T |
XP_006720633.1 |
XM_011521670.2 |
2061 |
Silent Mutation |
ACC,ACT |
T548T |
XP_011519972.1 |
XM_017022313.1 |
2061 |
Intron |
|
|
XP_016877802.1 |
View Full Product Details