Product Details

SNP ID
rs12594297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:72200343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCAATAAGGGAGAGGAACAGTC[A/G]CTGGGCCTTTTGCCCCACTAAGGTC
Phenotype
MIM: 179050
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRAMD2 PubMed Links

Gene Details

Gene
GRAMD2
Gene Name
GRAM domain containing 2
There are no transcripts associated with this gene.

Gene
PKM
Gene Name
pyruvate kinase, muscle
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206796.2 Intron NP_001193725.1
NM_001206797.2 Intron NP_001193726.1
NM_001206798.2 Intron NP_001193727.1
NM_001206799.1 Intron NP_001193728.1
NM_001316318.1 Intron NP_001303247.1
NM_002654.5 Intron NP_002645.3
NM_182470.3 Intron NP_872270.1
NM_182471.3 Intron NP_872271.1
XM_005254443.1 Intron XP_005254500.1
XM_005254445.4 Intron XP_005254502.1
XM_006720570.1 Intron XP_006720633.1
XM_011521670.2 Intron XP_011519972.1
XM_017022313.1 Intron XP_016877802.1

View Full Product Details