Product Details
- SNP ID
-
rs11139995
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:83244239 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGAGTCTCATGCAGCTCTCTCTGG[C/T]GTGGGAAGGCACCCTTTTCTGGCCA
- Phenotype
-
MIM: 607619
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FRMD3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75099214] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FRMD3
- Gene Name
- FERM domain containing 3
- Gene
- LOC102723989
- Gene Name
- uncharacterized LOC102723989
There are no transcripts associated with this gene.
View Full Product Details