Product Details

SNP ID
rs7343127
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57325950 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAGAGAAGGTTTTTTCTCTCTTT[A/G]GAGTGGCTGGTATAGGTCACCTGCC
Phenotype
MIM: 616847
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF543 PubMed Links

Gene Details

Gene
ZNF543
Gene Name
zinc finger protein 543
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213598.3 Intron NP_998763.2

View Full Product Details