Product Details

SNP ID
rs11881170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57660933 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGATCAGGATAAAATTGAGCCTTC[A/C]TTACTAGTTACTACGATTTTTGAAG
Phenotype
MIM: 613419
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZSCAN4 PubMed Links

Gene Details

Gene
ZSCAN4
Gene Name
zinc finger and SCAN domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152677.2 Intron NP_689890.1
XM_011526607.2 Intron XP_011524909.1
XM_017026458.1 Intron XP_016881947.1

View Full Product Details