Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_022496.4 | Intron | NP_071941.1 | |||
XM_017019821.1 | Intron | XP_016875310.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001319310.1 | Intron | NP_001306239.1 | |||
NM_001319311.1 | Intron | NP_001306240.1 | |||
NM_152317.3 | Intron | NP_689530.1 | |||
XM_011537876.2 | Intron | XP_011536178.1 | |||
XM_017018779.1 | Intron | XP_016874268.1 | |||
XM_017018780.1 | Intron | XP_016874269.1 | |||
XM_017018781.1 | Intron | XP_016874270.1 | |||
XM_017018782.1 | Intron | XP_016874271.1 | |||
XM_017018783.1 | Intron | XP_016874272.1 | |||
XM_017018784.1 | Intron | XP_016874273.1 | |||
XM_017018785.1 | Intron | XP_016874274.1 |