Product Details

SNP ID
rs13025223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218981496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGACAGGAGCAAATCTAGTACCA[G/T]ACAAGGATTGAGGGAGCTTCGGTCC
Phenotype
MIM: 600836 MIM: 607150
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CRYBA2 PubMed Links

Gene Details

Gene
CRYBA2
Gene Name
crystallin beta A2
There are no transcripts associated with this gene.

Gene
FEV
Gene Name
FEV, ETS transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017521.2 1470 UTR 3 NP_059991.1
Gene
LINC00608
Gene Name
long intergenic non-protein coding RNA 608
There are no transcripts associated with this gene.

View Full Product Details