Product Details

SNP ID
rs12262207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:69185273 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCATCCTCAAACTTAACATCAGC[C/G]TGGCTTCCTTAAGCATGAGGATCTG
Phenotype
MIM: 605122
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SUPV3L1 PubMed Links

Gene Details

Gene
SUPV3L1
Gene Name
Suv3 like RNA helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301683.1 Intron NP_001288612.1
NM_001323584.1 Intron NP_001310513.1
NM_001323585.1 Intron NP_001310514.1
NM_001323586.1 Intron NP_001310515.1
NM_001323587.1 Intron NP_001310516.1
NM_001323588.1 Intron NP_001310517.1
NM_003171.4 Intron NP_003162.2

View Full Product Details