Product Details
- SNP ID
-
rs12302465
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:50129746 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTGGAAAAGCTCTTAATTAGCTTA[C/T]GGATTCAGGAAACTGCGGATAGAGT
- Phenotype
-
MIM: 615335
MIM: 614478
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CERS5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79537743] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CERS5
- Gene Name
- ceramide synthase 5
- Gene
- COX14
- Gene Name
- COX14, cytochrome c oxidase assembly factor
There are no transcripts associated with this gene.
View Full Product Details