Product Details
- SNP ID
-
hCV30888919
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:21510282 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGGAGGCCGAGGAAAGCCTCCCC[C/G]ACTCCCCATTGTGCCTTATGACTCT
- Phenotype
-
MIM: 171760
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ALPL
PubMed Links
Gene Details
- Gene
- ALPL
- Gene Name
- alkaline phosphatase, liver/bone/kidney
There are no transcripts associated with this gene.
View Full Product Details