Product Details

SNP ID
rs12558511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:24490511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTTTTCTCTGAATCCTGAACTC[A/T]AAAAACTCTGAAAATTGAATTTCTT
Phenotype
MIM: 300906
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PDK3 PubMed Links

Gene Details

Gene
PDK3
Gene Name
pyruvate dehydrogenase kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142386.2 Intron NP_001135858.1
NM_005391.4 Intron NP_005382.1

View Full Product Details