Product Details
- SNP ID
-
rs7975939
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:7765864 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GATATTGAATTGGGAGTTTGACATA[C/G]AAGACTGGAAATAAAAATTTGAAAA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NANOGNB
PubMed Links
Gene Details
- Gene
- NANOGNB
- Gene Name
- NANOG neighbor homeobox
View Full Product Details