Product Details

SNP ID
rs11638588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74998182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTTTCATGAACAAAATTCAGGTA[G/T]TAACTGCTATCTTATTGATAATTAA
Phenotype
MIM: 613766
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SCAMP5 PubMed Links

Gene Details

Gene
SCAMP5
Gene Name
secretory carrier membrane protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178111.1 Intron NP_001171582.1
NM_001178112.1 Intron NP_001171583.1
NM_138967.3 Intron NP_620417.1
XM_006720420.1 Intron XP_006720483.1
XM_006720421.1 Intron XP_006720484.1
XM_006720422.3 Intron XP_006720485.1
XM_017021989.1 Intron XP_016877478.1

View Full Product Details