Product Details

SNP ID
rs12059107
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:232951606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCAGCTCCTTTGCAAGGCAAAT[C/T]TAATTTTGAAGTTTAGTGTGGGTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NTPCR PubMed Links

Gene Details

Gene
NTPCR
Gene Name
nucleoside-triphosphatase, cancer-related
There are no transcripts associated with this gene.

View Full Product Details