Product Details

SNP ID
rs12945355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76270379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCCCGCCCAGCCACCAGGCGCC[A/C]TGGATCCCGCCCTCCTCCCCCACCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
QRICH2 PubMed Links

Gene Details

Gene
QRICH2
Gene Name
glutamine rich 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032134.2 670 Intron NP_115510.1
XM_005257728.3 670 Intron XP_005257785.1
XM_006722136.3 670 Intron XP_006722199.1
XM_011525344.2 670 Intron XP_011523646.1
XM_017025206.1 670 Intron XP_016880695.1
XM_017025207.1 670 Intron XP_016880696.1
XM_017025208.1 670 Intron XP_016880697.1
Gene
UBALD2
Gene Name
UBA like domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182565.3 670 Silent Mutation CCA,CCC P123P NP_872371.1

View Full Product Details