Product Details

SNP ID
rs6851814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:151425285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGTAGCCAAATTCAGAGTGTAG[A/G]AATTTCTACAGGACAGATTACTCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM160A1 PubMed Links

Gene Details

Gene
FAM160A1
Gene Name
family with sequence similarity 160 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109977.1 Intron NP_001103447.1
XM_005263199.1 Intron XP_005263256.1
XM_011532220.1 Intron XP_011530522.1
XM_011532221.2 Intron XP_011530523.1
XM_011532222.2 Intron XP_011530524.1
XM_011532223.2 Intron XP_011530525.1
XM_011532224.2 Intron XP_011530526.1
XM_017008583.1 Intron XP_016864072.1

View Full Product Details