Product Details

SNP ID
rs11057971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:125002251 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAGTCCAGTTGCTGGGTCATACA[C/G]GAACTTTTTGTTTAACACTTTGGGG
Phenotype
MIM: 615627
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BRI3BP PubMed Links

Gene Details

Gene
BRI3BP
Gene Name
BRI3 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080626.5 Intron NP_542193.3
XM_011537940.2 Intron XP_011536242.1

View Full Product Details